1-[1-(5-Chloro-2-pyridinyl)-5-methyl-1H-pyrazol-4-yl]-1-ethanone ≥95%

Supplier: Matrix Scientific

047361-500MG 047361-5G
101879-660EA 278.46 USD
101879-660 101879-664
1-[1-(5-Chloro-2-pyridinyl)-5-methyl-1H-pyrazol-4-yl]-1-ethanone ≥95%
1-[1-(5-Chloro-2-pyridinyl)-5-methyl-1H-pyrazol-4-yl]-1-ethanone

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃O
Melting Pt: 138…140 °C
MDL Number: MFCD11100234
CAS Number: 210992-31-7

Order Now

Specification Test Results

Melting point 138-140°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR