Ethyl-5-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylate ≥95%

Supplier: Matrix Scientific

047354-500MG 047354-5G
101879-636EA 747.69 USD
101879-636 101879-640
Ethyl-5-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylate ≥95%
Ethyl-5-[(tert-butoxycarbonyl)amino]-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₉NO₅
Melting Pt: 121…123 °C
MDL Number: MFCD11840995
CAS Number: 1135283-35-0

Order Now

Specification Test Results

Melting point 121-123°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR