1-Methyl-1H-indazol-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-1-methylindazole, 1-methyl-1H-indazole-3-ylamine, 3-Amino-1-methyl-1H-indazole

047343-500MG 047343-5G
101879-594EA 27.69 USD
101879-594 101879-598
1-Methyl-1H-indazol-3-amine ≥95%
1-Methyl-1H-indazol-3-amine

Hazard: Irritant

Formula: C₈H₉N₃
MDL Number: MFCD09054755
CAS Number: 60301-20-4

Order Now

Specification Test Results

Melting point 85-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR