4-Chloro-1-benzothiophene-2-carbohydrazide ≥95%
Supplier: Matrix Scientific
047338-500MG
047338-5G
101879-574EA
278.46
USD
101879-574
101879-578
4-Chloro-1-benzothiophene-2-carbohydrazide ≥95%
4-Chloro-1-benzothiophene-2-carbohydrazide
Hazard: Irritant
Formula:
C₉H₇ClN₂OS MW: 226.69 g/mol Melting Pt: 184…186 °C |
MDL Number:
MFCD09817458 CAS Number: 952182-46-6 |
Specification Test Results
Melting point | 184-186°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...