2-(Methylsulfonyl)-4'-nitroacetophenone ≥95%
Supplier: Matrix Scientific
047332-500MG
047332-5G
101879-542EA
181.54
USD
101879-542
101879-546
2-(Methylsulfonyl)-4'-nitroacetophenone ≥95%
2-(Methylsulphonyl)-4'-nitroacetophenone
Hazard: Irritant
Formula:
C₉H₉NO₅S Melting Pt: 160-167°C |
MDL Number:
MFCD09865011 CAS Number: 136819-50-6 |
Specification Test Results
Melting point | 160-167°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...