1-(4-Methylphenyl)-2-oxo-3-pyrrolidinecarbonitrile ≥95%
Supplier: Matrix Scientific
047329-500MG
047329-5G
101879-526EA
278.46
USD
101879-526
101879-530
1-(4-Methylphenyl)-2-oxo-3-pyrrolidinecarbonitrile ≥95%
1-(4-Methylphenyl)-2-oxo-3-pyrrolidinecarbonitrile
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O MW: 200.24 g/mol Melting Pt: 132…134 °C |
MDL Number:
MFCD09817454 CAS Number: 930298-97-8 |
Specification Test Results
Melting point | 132-134°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...