4-Chlorobenzo[b]thiophene-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
047319-500MG
047319-5G
101879-478EA
278.46
USD
101879-478
101879-482
4-Chlorobenzo[b]thiophene-2-carboxylic acid ≥95%
4-Chlorobenzo[b]thiophene-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₅ClO₂S MW: 212.65 g/mol Melting Pt: 220-223°C |
MDL Number:
MFCD06655155 CAS Number: 23967-57-9 |
Specification Test Results
Melting point | 220-223°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...