Methyl-6-chlorobenzo[b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-chlorobenzo[b]thiophene-2-carboxylate
047310-500MG
047310-5G
101879-448EA
160
USD
101879-448
101879-452
Methyl-6-chlorobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 6-chlorobenzo[b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇ClO₂S MW: 226.6800 g/mol Melting Pt: 107-109°C |
MDL Number:
MFCD09027103 CAS Number: 104795-85-9 |
Specification Test Results
Melting point | 107-109°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...