7-Fluoro-1-benzothiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

047309-1G 047309-25G
101879-444EA 218.46 USD
101879-444 102551-370
7-Fluoro-1-benzothiophene-2-carboxylic acid ≥95%
7-Fluoro-1-benzothiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₅FO₂S
Melting Pt: 255…258 °C
MDL Number: MFCD07377074
CAS Number: 550998-67-9

Order Now

Specification Test Results

Melting point 255-258°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR