5-Fluorobenzo[b]thiophene-2-carboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Fluoro-1-benzothiophene-2-carboxylic acid
047306-5G
047306-25G
101879-430EA
260
USD
101879-430
102551-368
5-Fluorobenzo[b]thiophene-2-carboxylic acid ≥95%
5-Fluorobenzo[b]thiophene-2-carboxylic acid
Hazard: Irritant
Formula:
C₉H₅FO₂S MW: 196.19 g/mol Melting Pt: 240-243°C |
MDL Number:
MFCD01927184 CAS Number: 70060-13-8 |
Specification Test Results
Melting point | 240-243°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...