Methyl-7-fluorobenzo[b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
047305-500MG
047305-5G
101879-420EA
278.46
USD
101879-420
101879-424
Methyl-7-fluorobenzo[b]thiophene-2-carboxylate ≥95%
Methyl 7-fluorobenzo[b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇FO₂S MW: 210.224 g/mol Melting Pt: 84-87°C |
MDL Number:
MFCD09027102 CAS Number: 550998-54-4 |
Specification Test Results
Melting point | 84-87°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...