4'-{3-[4-(Benzyloxy)phenoxy]propoxy}acetophenone ≥95%
Supplier: Matrix Scientific
047303-500MG
047303-5G
101879-412EA
375.38
USD
101879-412
101879-416
4'-{3-[4-(Benzyloxy)phenoxy]propoxy}acetophenone ≥95%
4'-{3-[4-(Benzyloxy)phenoxy]propoxy}acetophenone
Hazard: Irritant
Formula:
C₂₄H₂₄O₄ Melting Pt: 76…78 °C |
MDL Number:
MFCD08689786 CAS Number: 937602-11-4 |
Specification Test Results
Melting point | 76-78°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...