Methyl-4-[3-(3-formylphenoxy)propoxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
047295-500MG
047295-5G
101879-364EA
375.38
USD
101879-364
101879-368
Methyl-4-[3-(3-formylphenoxy)propoxy]benzenecarboxylate ≥95%
Methyl-4-[3-(3-formylphenoxy)propoxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₈H₁₈O₅ MW: 314.34 g/mol Melting Pt: 71…73 °C |
MDL Number:
MFCD08689779 CAS Number: 937601-98-4 |
Specification Test Results
Melting point | 71-73°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...