5-Methyl-2,1,3-benzothiadiazole ≥95%
Supplier: Matrix Scientific
047293-500MG
047293-5G
101879-352EA
72.31
USD
101879-352
101879-356
5-Methyl-2,1,3-benzothiadiazole ≥95%
5-Methyl-2,1,3-benzothiadiazole
Hazard: Irritant
Formula:
C₇H₆N₂S Melting Pt: 31-32°C |
MDL Number:
MFCD00173945 CAS Number: 1457-93-8 |
Specification Test Results
Melting point | 31-32°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...