Methyl-3-[2-(3-acetylphenoxy)ethoxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
047289-500MG
047289-5G
101879-328EA
375.38
USD
101879-328
101879-332
Methyl-3-[2-(3-acetylphenoxy)ethoxy]benzenecarboxylate ≥95%
Methyl-3-[2-(3-acetylphenoxy)ethoxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₁₈H₁₈O₅ MW: 314.34 g/mol Melting Pt: 88…90 °C |
MDL Number:
MFCD08689775 CAS Number: 937601-96-2 |
Specification Test Results
Melting point | 88-90°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...