Methyl-3-[2-(3-formylphenoxy)ethoxy]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

047287-500MG 047287-5G
101879-316EA 375.38 USD
101879-316 101879-320
Methyl-3-[2-(3-formylphenoxy)ethoxy]benzenecarboxylate ≥95%
Methyl-3-[2-(3-formylphenoxy)ethoxy]benzenecarboxylate

Hazard: Irritant

Formula: C₁₇H₁₆O₅
MW: 300.31 g/mol
Melting Pt: 90…92 °C
MDL Number: MFCD08689773
CAS Number: 937601-94-0

Order Now

Specification Test Results

Melting point 90-92°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR