Methyl-4-[2-(4-benzylpiperazino)ethoxy]benzenecarboxylate ≥95%
Supplier: Matrix Scientific
047284-500MG
047284-5G
101879-300EA
278.46
USD
101879-300
101879-304
Methyl-4-[2-(4-benzylpiperazino)ethoxy]benzenecarboxylate ≥95%
Methyl-4-[2-(4-benzylpiperazino)ethoxy]benzenecarboxylate
Hazard: Irritant
Formula:
C₂₁H₂₆N₂O₃ MW: 354.45 g/mol Melting Pt: 58…60 °C |
MDL Number:
MFCD08689770 CAS Number: 937601-91-7 |
Specification Test Results
Melting point | 58-60°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...