3,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide ≥95%

Supplier: Matrix Scientific

047280-500MG 047280-5G
101879-276EA 161.54 USD
101879-276 101879-280
3,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide ≥95%
3,4-Dichloro-N-(2-hydroxyethyl)benzenecarboxamide

Hazard: Irritant

Formula: C₉H₉Cl₂NO₂
Melting Pt: 153…155 °C
MDL Number: MFCD00461856
CAS Number: 28298-26-2

Order Now

Specification Test Results

Melting point 153-155°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR