4'-{2-[4-(Benzyloxy)phenoxy]ethoxy}acetophenone ≥95%

Supplier: Matrix Scientific

047278-500MG 047278-5G
101879-264EA 278.46 USD
101879-264 101879-268
4'-{2-[4-(Benzyloxy)phenoxy]ethoxy}acetophenone ≥95%
4'-{2-[4-(Benzyloxy)phenoxy]ethoxy}acetophenone

Hazard: Irritant

Formula: C₂₃H₂₂O₄
Melting Pt: 170…172 °C
MDL Number: MFCD08689767
CAS Number: 937601-89-3

Order Now

Specification Test Results

Melting point 170-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR