4-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

047276-500MG 047276-5G
101879-252EA 278.46 USD
101879-252 101879-256
4-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde ≥95%
4-{2-[4-(Benzyloxy)phenoxy]ethoxy}benzenecarbaldehyde

Hazard: Irritant

Formula: C₂₂H₂₀O₄
MW: 348.4 g/mol
Melting Pt: 123…125 °C
MDL Number: MFCD08689765
CAS Number: 937601-87-1

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR