Methyl-4-[2-(4-acetylphenoxy)ethoxy]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

047274-500MG 047274-5G
101879-240EA 278.46 USD
101879-240 101879-244
Methyl-4-[2-(4-acetylphenoxy)ethoxy]benzenecarboxylate ≥95%
Methyl-4-[2-(4-acetylphenoxy)ethoxy]benzenecarboxylate

Hazard: Irritant

Formula: C₁₈H₁₈O₅
MW: 314.34 g/mol
Melting Pt: 134…136 °C
MDL Number: MFCD08689763
CAS Number: 937601-85-9

Order Now

Specification Test Results

Melting point 134-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR