6-(Methylamino)nicotinonitrile ≥95%

Supplier: Matrix Scientific

047259-500MG 047259-5G
101879-160EA 278.46 USD
101879-160 101879-164
6-(Methylamino)nicotinonitrile ≥95%
6-(Methylamino)nicotinonitrile

Hazard: Irritant

Formula: C₇H₇N₃
Melting Pt: 131…133 °C
MDL Number: MFCD08689756
CAS Number: 261715-36-0

Order Now

Specification Test Results

Melting point 131-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR