N-Methyl-5-nitro-2-pyridinamine ≥95%
Supplier: Matrix Scientific
047253-500MG
047253-5G
101879-124EA
181.54
USD
101879-124
101879-128
N-Methyl-5-nitro-2-pyridinamine ≥95%
N-Methyl-5-nitro-2-pyridinamine
Hazard: Irritant
Formula:
C₆H₇N₃O₂ Melting Pt: 178…180 °C |
MDL Number:
MFCD00152624 CAS Number: 4093-89-4 |
Specification Test Results
Melting point | 178-180°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...