3-(2,5-Dichlorophenyl)acrylic acid ≥95%

Supplier: Matrix Scientific

047248-500MG 047248-5G
101879-098EA 181.54 USD
101879-098 101879-102
3-(2,5-Dichlorophenyl)acrylic acid ≥95%
3-(2,5-Dichlorophenyl)acrylic acid

Hazard: Irritant

Formula: C₉H₆Cl₂O₂
Melting Pt: 192…194 °C
MDL Number: MFCD03002786
CAS Number: 101869-82-3

Order Now

Specification Test Results

Melting point 192-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR