4-[4-(Hydroxymethyl)piperidino]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-[4-(Hydroxymethyl)piperidin-1-yl]benzonitrile

047244-500MG 047244-5G
101879-082EA 278.46 USD
101879-082 101879-086
4-[4-(Hydroxymethyl)piperidino]benzenecarbonitrile ≥95%
4-[4-(Hydroxymethyl)piperidino]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₁₆N₂O
MW: 216.29 g/mol
Melting Pt: 149…151 °C
MDL Number: MFCD08689746
CAS Number: 162997-46-8

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR