{1-[(4-Chlorophenyl)sulfonyl]-4-piperidinyl}methanol ≥95%
Supplier: Matrix Scientific
047237-500MG
047237-5G
101879-052EA
278.46
USD
101879-052
101879-056
{1-[(4-Chlorophenyl)sulfonyl]-4-piperidinyl}methanol ≥95%
{1-[(4-Chlorophenyl)sulfonyl]-4-piperidinyl}methanol
Hazard: Irritant
Formula:
C₁₂H₁₆ClNO₃S MW: 289.78 g/mol Melting Pt: 102…104 °C |
MDL Number:
MFCD08689742 CAS Number: 870529-81-0 |
Specification Test Results
Melting point | 102-104°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...