Dimethyl-4-iodo-1H-pyrazole-3,5-dicarboxylate ≥95%

Supplier: Matrix Scientific

047235-500MG 047235-5G
101879-040EA 153.85 USD
101879-040 101879-044
Dimethyl-4-iodo-1H-pyrazole-3,5-dicarboxylate ≥95%
Dimethyl 4-iodo-1H-pyrazole-3,5-dicarboxylate

Hazard: Irritant

Formula: C₇H₇IN₂O₄
Melting Pt: 176…178 °C
MDL Number: MFCD08689740
CAS Number: 1027819-68-6

Order Now

Specification Test Results

Melting point 176-178°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR