3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Ethyl-2-thioxo-2,3-dihydroquinazolin-4(1H)-one

047233-500MG 047233-5G
101879-028EA 181.54 USD
101879-028 101879-032
3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone ≥95%
3-Ethyl-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

Hazard: Irritant

Formula: C₁₀H₁₀N₂OS
MW: 206.27 g/mol
Melting Pt: 256…258 °C
MDL Number: MFCD00126093
CAS Number: 13906-08-6

Order Now

Specification Test Results

Melting point 256-258°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR