Phenyl-N-[2-(1-pyrrolidinyl)ethyl]carbamate hydrochloride ≥95%

Supplier: Matrix Scientific

047221-500MG 047221-5G
101878-988EA 278.46 USD
101878-988 101878-992
Phenyl-N-[2-(1-pyrrolidinyl)ethyl]carbamate hydrochloride ≥95%
Phenyl-N-[2-(1-pyrrolidinyl)ethyl]carbamate hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
Melting Pt: 154…156 °C
MDL Number: MFCD09027099
CAS Number: 1170129-51-7

Order Now

Specification Test Results

Melting point 154-156°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR