Phenyl-N-(2-morpholinoethyl)carbamate hydrochloride ≥95%
Supplier: Matrix Scientific
047219-500MG
047219-5G
101878-976EA
181.54
USD
101878-976
101878-980
Phenyl-N-(2-morpholinoethyl)carbamate hydrochloride ≥95%
Phenyl-N-(2-morpholinoethyl)carbamate hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₃ MW: 286.76 g/mol Melting Pt: 160…162 °C |
MDL Number:
MFCD09027097 CAS Number: 1171158-20-5 |
Specification Test Results
Melting point | 160-162°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...