(S)-(-)-Stericol ≥99%

Supplier: Matrix Scientific

Synonyms: (S)-(−)-1-(2,4,6-Triisopropylphenyl)ethanol

047215-500MG 047215-5G
101878-956EA 278.46 USD
101878-956 101878-960
(S)-(-)-Stericol ≥99%
(S)-(-)-Stericol

Hazard: Irritant

Formula: C₁₇H₂₈O
MDL Number: MFCD09836213
CAS Number: 102225-88-7

Order Now

Specification Test Results

Melting point 88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR