Ethyl-5-(2-quinolinyl)-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
047205-500MG
047205-1G
047205-5G
101878-918EA
375.38
USD
101878-918
101878-920
101878-922
Ethyl-5-(2-quinolinyl)-2-thiophenecarboxylate ≥95%
Ethyl-5-(2-quinolinyl)-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₁₆H₁₃NO₂S Melting Pt: 62…64 °C |
MDL Number:
MFCD12498718 CAS Number: 1171919-15-5 |
Specification Test Results
Melting point | 62-64°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...