Methyl-5-iodo-2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 5-iodo-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
047202-500MG
047202-1G
047202-5G
101878-904EA
556.92
USD
101878-904
101878-906
101878-908
Methyl-5-iodo-2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate ≥95%
Methyl-5-iodo-2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylate
Hazard: Irritant
Formula:
C₆H₅IN₂O₄ MW: 296.02 g/mol Melting Pt: 283…285 °C |
MDL Number:
MFCD12498716 CAS Number: 116393-71-6 |
Specification Test Results
Melting point | 283-285°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...