2-(Methylsulfanyl)-6-(trifluoromethyl)-4-pyrimidinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-(methylthio)-6-(trifluoromethyl)pyrimidine

047198-500MG 047198-1G 047198-5G
101878-888EA 921.54 USD
101878-888 101878-890 101878-892
2-(Methylsulfanyl)-6-(trifluoromethyl)-4-pyrimidinamine ≥95%
2-(Methylsulfanyl)-6-(trifluoromethyl)-4-pyrimidinamine

Hazard: Irritant

Formula: C₆H₆F₃N₃S
MW: 209.19 g/mol
Melting Pt: 120…121 °C
MDL Number: MFCD01011766
CAS Number: 85730-32-1

Order Now

Specification Test Results

Melting point 120-121°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR