Ethyl-3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Supplier: Matrix Scientific
047194-500MG
047194-1G
101878-872EA
556.92
USD
101878-872
101878-874
Ethyl-3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate ≥95%
Ethyl 3-nitro-6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₈F₃N₃O₄ Melting Pt: 134…135 °C |
MDL Number:
MFCD12498712 CAS Number: 1171919-07-5 |
Specification Test Results
Melting point | 134-135°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...