3-Bromo-4-chloro-7-azaindole ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-4-chloro-1H-pyrrolo[2,3-b]pyridine
047193-500MG
047193-1G
101878-868EA
747.69
USD
101878-868
101878-870
3-Bromo-4-chloro-7-azaindole ≥95%
3-Bromo-4-chloro-7-azaindole
Hazard: Irritant
Formula:
C7H4BrClN2 MW: 231.4800 g/mol Melting Pt: 214…215 °C |
MDL Number:
MFCD09880132 CAS Number: 1000340-39-5 |
Specification Test Results
Melting point | 214-215°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...