4-Iodopyridine-2-carboxylic acid hydroiodide ≥95%

Supplier: Matrix Scientific

047191-500MG 047191-1G
101878-860EA 261.54 USD
101878-860 101878-862
4-Iodopyridine-2-carboxylic acid hydroiodide ≥95%
4-Iodopyridine-2-carboxylic acid hydroiodide

Hazard: Irritant

Formula: C₆H₅I₂NO₂
Melting Pt: 202…204 °C
MDL Number: MFCD06738838
CAS Number: 959771-81-4

Order Now

Specification Test Results

Melting point 202-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR