6-Chloro-2-phenyl-4-pyrimidinyl-4-methoxybenzyl ether ≥95%
Supplier: Matrix Scientific
047188-500MG
047188-1G
047188-5G
101878-846EA
278.46
USD
101878-846
101878-848
101878-850
6-Chloro-2-phenyl-4-pyrimidinyl-4-methoxybenzyl ether ≥95%
6-Chloro-2-phenyl-4-pyrimidinyl-4-methoxybenzyl ether
Hazard: Irritant
Formula:
C₁₈H₁₅ClN₂O₂ MW: 326.79 g/mol Melting Pt: 94…96 °C |
MDL Number:
MFCD12498709 CAS Number: 1044559-74-1 |
Specification Test Results
Melting point | 94-96°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...