2-Chloro-3,5-diiodo-4-pyridinamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-3,5-diiodopyridin-4-amine

047186-500MG 047186-1G 047186-5G
101878-834EA 375.38 USD
101878-834 101878-836 101878-838
2-Chloro-3,5-diiodo-4-pyridinamine ≥95%
2-Chloro-3,5-diiodo-4-pyridinamine

Hazard: Irritant

Formula: C₅H₃ClI₂N₂
MW: 380.35 g/mol
Melting Pt: 158…159 °C
MDL Number: MFCD12498707
CAS Number: 1171919-00-8

Order Now

Specification Test Results

Melting point 158-159°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR