3-(Trifluoromethyl)-5-isoxazolamine ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Amino-3-(trifluoromethyl)isoxazole
047185-500MG
047185-1G
101878-830EA
150.77
USD
101878-830
101878-832
3-(Trifluoromethyl)-5-isoxazolamine ≥95%
3-(Trifluoromethyl)-5-isoxazolamine
Hazard: Irritant
Formula:
C₄H₃F₃N₂O MW: 152.08 g/mol Melting Pt: 53…55 °C |
MDL Number:
MFCD04972407 CAS Number: 108655-63-6 |
Specification Test Results
Melting point | 53-55°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...