Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate ≥95%
Supplier: Matrix Scientific
047184-500MG
047184-1G
047184-5G
101878-824EA
300
USD
101878-824
101878-826
101878-828
Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate ≥95%
Ethyl-4,6-dihydroxy-2-(trifluoromethyl)nicotinate
Hazard: Irritant
Formula:
C₉H₈F₃NO₄ MW: 251.16 g/mol Melting Pt: 217…219 °C |
MDL Number:
MFCD09881353 CAS Number: 103900-77-2 |
Specification Test Results
Melting point | 217-219°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...