tert-Butyl-N-{3-[(tert-butoxycarbonyl)amino]-2-hydroxypropyl}carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: Di-tert-butyl (2-hydroxypropane-1,3-diyl)dicarbamate

047182-500MG 047182-1G 047182-5G
101878-814EA 295.38 USD
101878-814 101878-816 101878-818
tert-Butyl-N-{3-[(tert-butoxycarbonyl)amino]-2-hydroxypropyl}carbamate ≥95%
tert-Butyl-N-{3-[(tert-butoxycarbonyl)amino]-2-hydroxypropyl}carbamate

Hazard: Irritant

Formula: C₁₃H₂₆N₂O₅
MW: 290.36 g/mol
Melting Pt: 99…102 °C
MDL Number: MFCD12498706
CAS Number: 98642-15-0

Order Now

Specification Test Results

Melting point 99-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR