8-Chloro-6-(trifluoromethyl)-4(1H)-quinolinone ≥95%
Supplier: Matrix Scientific
047177-500MG
047177-1G
047177-5G
101878-790EA
375.38
USD
101878-790
101878-792
101878-794
8-Chloro-6-(trifluoromethyl)-4(1H)-quinolinone ≥95%
8-Chloro-6-(trifluoromethyl)-4(1H)-quinolinone
Hazard: Irritant
Formula:
C₁₀H₅ClF₃NO MW: 247.61 g/mol Melting Pt: 285…286 °C |
MDL Number:
MFCD12827776 CAS Number: 1065092-49-0 |
Specification Test Results
Melting point | 285-286°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...