2-(tert-Butyl)-4,5-dichloro-3(2H)-pyridazinone ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(tert-Butyl)-4,5-dichloropyridazin-3(2H)-one
047176-500MG
047176-1G
047176-5G
101878-784EA
430.77
USD
101878-784
101878-786
101878-788
2-(tert-Butyl)-4,5-dichloro-3(2H)-pyridazinone ≥95%
2-(tert-Butyl)-4,5-dichloro-3(2H)-pyridazinone
Hazard: Irritant
Formula:
C₈H₁₀Cl₂N₂O MW: 221.09 g/mol Melting Pt: 61…62 °C |
MDL Number:
MFCD00277876 CAS Number: 84956-71-8 |
Specification Test Results
Melting point | 61-62°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...