Ethyl-5-amino-1-[4-bromo-3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

047175-500MG 047175-1G
101878-780EA 556.92 USD
101878-780 101878-782
Ethyl-5-amino-1-[4-bromo-3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylate ≥95%
Ethyl-5-amino-1-[4-bromo-3-(trifluoromethyl)phenyl]-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₁BrF₃N₃O₂
Melting Pt: 151…152 °C
MDL Number: MFCD12827774
CAS Number: 1186404-53-4

Order Now

Specification Test Results

Melting point 151-152°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR