Ethyl-5-amino-1-(2,4-difluorophenyl)-1H-pyrazole-4-carboxylate ≥95%

Supplier: Matrix Scientific

047173-500MG 047173-1G 047173-5G
101878-770EA 375.38 USD
101878-770 101878-772 101878-774
Ethyl-5-amino-1-(2,4-difluorophenyl)-1H-pyrazole-4-carboxylate ≥95%
Ethyl 5-amino-1-(2,4-difluorophenyl)-1H-pyrazole-4-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₁F₂N₃O₂
Melting Pt: 127…128 °C
MDL Number: MFCD03407822
CAS Number: 138907-72-9

Order Now

Specification Test Results

Melting point 127-128°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR