1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbonitrile ≥95%
Supplier: Matrix Scientific
047170-500MG
047170-1G
047170-5G
101878-756EA
278.46
USD
101878-756
101878-758
101878-760
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbonitrile ≥95%
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarbonitrile
Hazard: Irritant
Formula:
C₁₂H₁₁ClF₃N₃ Melting Pt: 108…109 °C |
MDL Number:
MFCD12498704 CAS Number: 1171918-97-0 |
Specification Test Results
Melting point | 108-109°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...