4,6-Dichloro-8-iodoquinoline ≥95%

Supplier: Matrix Scientific

047168-500MG 047168-1G
101878-750EA 747.69 USD
101878-750 101878-752
4,6-Dichloro-8-iodoquinoline ≥95%
4,6-Dichloro-8-iodoquinoline

Hazard: Irritant

Formula: C₉H₅Cl₂N
Melting Pt: 134…136 °C
MDL Number: MFCD12498702
CAS Number: 1171918-94-7

Order Now

Specification Test Results

Melting point 134-136°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR