2-(Hydroxymethyl)-5-methoxy-4H-pyran-4-one ≥95%

Supplier: Matrix Scientific

047167-500MG 047167-1G
101878-746EA 375.38 USD
101878-746 101878-748
2-(Hydroxymethyl)-5-methoxy-4H-pyran-4-one ≥95%
2-(Hydroxymethyl)-5-methoxy-4H-pyran-4-one

Hazard: Irritant

Formula: C₇H₈O₄
Melting Pt: 165…167 °C
MDL Number: MFCD00224874
CAS Number: 6269-25-6

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR