3-Iodo-5-methoxy-6-(trifluoromethyl)-1H-indole ≥95%

Supplier: Matrix Scientific

047165-500MG 047165-1G
101878-742EA 921.54 USD
101878-742 101878-744
3-Iodo-5-methoxy-6-(trifluoromethyl)-1H-indole ≥95%
3-Iodo-5-methoxy-6-(trifluoromethyl)-1H-indole

Hazard: Irritant

Formula: C₁₀H₇F₃INO
Melting Pt: 130 °C
MDL Number: MFCD12498701
CAS Number: 1171918-93-6

Order Now

Specification Test Results

Melting point 130°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR